Learn More
Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals
30.65 € - 91.76 €
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357608
|
Thermo Scientific Alfa Aesar
033295.22 |
100 g |
30.65 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347608
|
Thermo Scientific Alfa Aesar
033295.36 |
500 g |
91.76 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium carbonate is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles show a useful application as an anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
800°C (decomposition) | |
White | |
99.0% min. | |
MFCD00010906 | |
Soluble in acids. Insoluble in water. | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
ACS | |
White powder | |
Calcium carbonate, For ACS analysis |
471-34-1 | |
Powder | |
CCaO3 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
1.6583 | |
2.93 |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
missing translation for 'rtecsNumber' : FF9335000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.