missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, ACS, low in alkalies, 99.0% min, Thermo Scientific Chemicals

58.09 € - 155.78 €

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383418
View Documents
Thermo Scientific Alfa Aesar
036337.30
250 g
58.09 €
250g
Please to purchase this item. Need a web account? Register with us today!
11373418
View Documents
Thermo Scientific Alfa Aesar
036337.A1
1 kg
155.78 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium carbonate
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311
[Ca++].[O-]C([O-])=O

Specifications

800°C (decomposition)
White
99.0% min.
MFCD00010906
Soluble in acids,insoluble in water.
[Ca++].[O-]C([O-])=O
100.09
CHEBI:3311
ACS
2.93
471-34-1
Powder
CCaO3
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
10112
100.09
1.6583
Calcium carbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 207-439-9

missing translation for 'rtecsNumber' : FF9335000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.