missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, ACS reagent, chelometric standard, Thermo Scientific Chemicals
37.39 € - 156.78 €
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15515096
|
Thermo Scientific Acros
403801000 |
100 g |
37.39 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15525096
|
Thermo Scientific Acros
403805000 |
500 g |
156.78 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
825.0°C | |
CCaO3 | |
MFCD00010906 | |
15,1659 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted acids | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
99.95 to 100.05% (on dried substance) (Complexometry) | |
Calcium carbonate |
471-34-1 | |
CaCO3 | |
01,103; 02,57; 04,67; 05,89 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
ACS Reagent |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
Product Identifier
- Calcium carbonate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title