missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate(CaCO3), Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| CAS | 471-34-1 |
| Quantity | 500 g |
| Molecular Formula | CCaO3 |
| Linear Formula | CaCO3 |
| MDL Number | MFCD00010906 |
| UN Number | NONH for all modes of transport |
| Beilstein | 8008338 |
| Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
| InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
| SMILES | [Ca++].[O-]C([O-])=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction