missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Powder, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
Specifications
Specifications
CAS | 471-34-1 |
Physical Form | Powder |
Quantity | 2.5 kg |
Molecular Formula | CCaO3 |
MDL Number | MFCD00010906 |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
SMILES | [Ca++].[O-]C([O-])=O |
IUPAC Name | calcium;carbonate |
Molecular Weight (g/mol) | 100.09 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction