missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15497485
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Applied as neutralizing agent, filler, and extender in rubber, plastics, paints, opacifying agent in paper, putty, antacid, Portland cement, sulfur dioxide, removal from stack gases, allurgical flux, analytical chemistry, carbon dioxide generation, and determining halogens in organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,1657
Solubility Information Soluble in dilute acids (5 M HCl: 0.1 M at 20°C,clear,colorless). Insoluble in water.
IUPAC Name calcium carbonate
Formula Weight 100.09
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Density 2.930 g/mL
Assay (metals basis)
Chemical Name or Material Calcium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.