missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals

53.67 € - 75.17 €

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445389
View Documents
Thermo Scientific Alfa Aesar
J63122.AD
50 mL
53.67 €
50mL
Please to purchase this item. Need a web account? Register with us today!
15455389
View Documents
Thermo Scientific Alfa Aesar
J63122.AE
100 mL
75.17 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong acids, borane/boron oxides, zinc, calcium oxide, methyl vinyl ether. Calcium chloride is attacked by bromine trifluoride. Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant.
Specifications

Chemical Identifiers

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
calcium dichloride
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312
[Cl-].[Cl-].[Ca++]

Specifications

10043-52-4
Liquid
MFCD00010903
Soluble in water (74.5g/100ml) at 20°C,alcohol,water (159g/100ml) at 100°C,acetic acid,and acetone.
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
1M aq. soln.
Calcium chloride
Colorless
CaCl2
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Calcium chloride, 1M aq. soln

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : calcium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only