missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 96%, for biochemistry, anhydrous, Thermo Scientific Chemicals

92.96 € - 293.46 €

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12674977
View Documents
Thermo Scientific Acros
192731000
100 g
92.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
12684977
View Documents
Thermo Scientific Acros
192735000
500 g
293.46 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10043-52-4
110.98
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312
[Cl-].[Cl-].[Ca++]
CaCl2
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
calcium dichloride

Specifications

>1600.0°C
10043-52-4
CaCl2
01,104; 04,368
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
Biochemical
Calcium chloride
782.0°C
95% min. (Argentometry)
CaCl2
15,1661
Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
96%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 233-140-8

Product Identifier
  • Calcium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Calcium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only