missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, ACS reagent, desiccant, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | >1600.0°C |
Melting Point | 782.0°C |
Quantity | 500 g |
Linear Formula | CaCl2 |
Fieser | 01,104; 04,368 |
Merck Index | 15,1661 |
Solubility Information | Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol |
IUPAC Name | calcium dichloride |
Formula Weight | 110.99 |
Grade | ACS Reagent |
Show More |
Safety and Handling
Product Identifier
- Calcium chloride
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Calcium chloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction