missing translation for 'onlineSavingsMsg'
Learn More
Calcium chloride, anhydrous, 93%, Thermo Scientific Chemicals Product Code.: 11301689

Calcium chloride, anhydrous, 93%, Thermo Scientific Chemicals

Product Code. 11301689
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11301689

Brand: Thermo Scientific Alfa Aesar 012316.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

It is used as a drying and dehydrating agent for organic liquids and gases, and for solids in desiccators. Common applications include brine for refrigeration plants, ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishders, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium, and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcalShow MoreShow Less
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point >1600°C
Melting Point 782°C
Color White to Gray
Physical Form Granular Powder
Quantity 1 kg
Assay Percent Range 93%
Solubility Information Freely soluble in water,ethanol,methanol,and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide,and ethyl acetate.
IUPAC Name calcium;dichloride
Formula Weight 110.99
Sensitivity Hygroscopic
Appearance White to gray granular powder
Density 3.08g/cm3
Chemical Name or Material Calcium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L023 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number012316.A7,012316.A1
Lot NumberP24L022 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number012316.A7,012316.A1
Lot NumberP17L024 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number012316.A7,012316.A1
Lot NumberP17L025 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number012316.A7,012316.A1
Lot NumberZ02K018 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number012316.A7,012316.A1
compliance-icons
Product Identifier
  • Calcium chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium chloride, anhydrous, 93%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.