missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99+%, ACS reagent

Product Code. 10158280 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10158280

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02
MDL Number MFCD00149613

Specifications

Melting Point 175.0°C
Color White
pH 4.5 to 8.5 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99.0 to 105% (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Formula Weight 147.02
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.