missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10403265
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02

Spezifikation

Melting Point 175.0°C
Color White
pH 4.5 to 6.5 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Formula Weight 147.02
Percent Purity ≥99%
Grade Analytical
Packaging Plastic Bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Calcium chloride dihydrate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
Gefahrenhinweis
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'