missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 4 x 2.5 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 147.01g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Vapor Pressure | 0.01mmHg (20°C) |
Chemical Name or Material | Calcium Chloride Dihydrate |
Safety and Handling
Product Identifier
- Calcium chloride dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'