missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, ACS Reagent, Honeywell Fluka™

Product Code. 15692220
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
MDL Number MFCD00149613

Specifications

Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS Reagent
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Calcium Chloride Dihydrate
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'