missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP
Specifications
Specifications
Melting Point | 175°C |
Color | Beige to White |
pH | 4.5 to 6.5 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 147.02g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Calcium chloride dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction