missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Dihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

188.00 € - 482.00 €

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15517904
View Documents
Avantor J.T.Baker
1332-01
500 g Poly Bottle
188.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
15527904
View Documents
Avantor J.T.Baker
1332-05
2.5 kg Poly Bottle
482.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

10035-04-8
CaCl2·2H2O

Specifications

10035-04-8
CaCl2·2H2O
BAKER ANALYZED™ ACS Reagent
Calcium Chloride, Dihydrate
Granules
147.01
1L = 0.84kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 500GR CALCIUMCHLORIDE 2AQ BAKER ANALYZED A.C.S. Re

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Calcium chloride, dihydrate
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.