missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, puriss., Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
15 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 2.5 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 147.01g/mol |
Percent Purity | 99 to 103% |
Grade | puriss. |
Chemical Name or Material | Calcium Chloride Dihydrate, Meets analytical specification of Ph. Eur.USP, FCC |
Safety and Handling

Product Identifier
- Calcium chloride dihydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Calcium chloride dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction