missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Extra Pure, SLR, Fused, Granular, Fisher Chemical™
28.20 € - 88.30 €
Chemical Identifiers
CAS | 10043-52-4 |
---|---|
Molecular Formula | CaCl2 |
Molecular Weight (g/mol) | 110.98 |
MDL Number | 10903 |
InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
IUPAC Name | calcium dichloride |
SMILES | [Cl-].[Cl-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10161800
|
Fisher Chemical
C/1400/53 |
500 g |
28.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488093
|
Fisher Chemical
C/1400/60 |
1 kg |
45.55 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10478590
|
Fisher Chemical
C/1400/62 |
2.5 kg |
88.30 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium Chloride, Extra Pure, SLR, Fused, Granular
Chemical Identifiers
10043-52-4 | |
110.98 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
[Cl-].[Cl-].[Ca++] |
CaCl2 | |
10903 | |
calcium dichloride |
Specifications
1600°C | |
10043-52-4 | |
8 to 10 | |
CaCl2 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium dichloride | |
110.99g/mol |
782°C | |
Beige | |
Solid | |
10903 | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium chloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.