Learn More
Calcium chloride hexahydrate, 97+%, extra pure, Thermo Scientific Chemicals
31.86 € - 73.37 €
Chemical Identifiers
CAS | 7774-34-7 |
---|---|
Molecular Formula | CaCl2·6H2O |
Molecular Weight (g/mol) | 219.08 |
MDL Number | MFCD00149616 |
InChI Key | QHFQAJHNDKBRBO-UHFFFAOYSA-L |
Synonym | calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g |
PubChem CID | 6093252 |
IUPAC Name | calcium;dichloride;hexahydrate |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10610414
|
Thermo Scientific Acros
389250010 |
1 kg | Plastic bottle |
31.86 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542444
|
Thermo Scientific Acros
389250025 |
2.5 kg | Plastic bottle |
73.37 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7774-34-7 | |
219.08 | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
6093252 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] |
CaCl2·6H2O | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
calcium;dichloride;hexahydrate |
Specifications
30.0°C | |
Colorless | |
97.0 to 103% (Complexometry) | |
CaCl2·6H2O | |
15, 1661 | |
Solubility in water: 5360g/L (20°C) | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2] | |
219.08 | |
219.08 | |
Extra Pure |
7774-34-7 | |
Chunks | |
CaCl2·6H2O | |
MFCD00149616 | |
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g | |
QHFQAJHNDKBRBO-UHFFFAOYSA-L | |
calcium;dichloride;hexahydrate | |
6093252 | |
≥97% | |
Calcium chloride hexahydrate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Calcium chloride hexahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.