missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride solution, Fisher Chemical™

Product Code. 12920924
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 10043-52-4
Quantity 1000 mL
Molecular Formula CaCl2
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ca++]
IUPAC Name calcium;dichloride
Molecular Weight (g/mol) 110.98
PubChem CID 5284359
ChEBI CHEBI:3312
Concentration or Composition (by Analyte or Components) 0.5M (1N)
Packaging Bottle
Chemical Name or Material Calcium Chloride Solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'