missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Technical, Fused, Porous Lump, Fisher Chemical™

Product Code. 10630341
Click to view available options
Quantity:
1 kg
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number 10903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point 1600°C
Melting Point 782°C
Color Beige
pH 8 to 10
Physical Form Solid
Quantity 1 kg
Formula Weight 110.99g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Calcium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.