missing translation for 'onlineSavingsMsg'
Learn More

Calcium crystalline dendritic pieces, 99.98% (metals basis), Thermo Scientific Chemicals

Product Code. 11382408
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

UN Number UN1401
Quantity 5 g
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Form Pieces
Assay (metals basis)
Chemical Name or Material Calcium crystalline dendritic pieces

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'