missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11415300
Click to view available options
Quantity:
30 g
60 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.372
MDL Number MFCD00064209
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
PubChem CID 9290
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Specifications

Quantity 30 g
Solubility Information Slowly soluble in water. Insoluble in alcohol,acetic acid and organic solvents.
Formula Weight 430.37
Concentration or Composition (by Analyte or Components) 2.5% w/w aq. soln.
Physical Form Gel
Chemical Name or Material Calcium D-gluconate gel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.