missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium fluoride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
Solubility
Soluble in ammonium salts. Soluble in water (slightly).
Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
Solubility
Soluble in ammonium salts. Soluble in water (slightly).
Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
Specifications
Specifications
Boiling Point | ∼2500°C |
Melting Point | 1403°C |
Quantity | 250 g |
Merck Index | 14,1667 |
Solubility Information | Soluble in ammonium salts. Soluble in water (slightly). |
Formula Weight | 78.08 |
Percent Purity | 97% |
Odor | Odorless |
Refractive Index | 1.434 |
Density | 3.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'