missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals

57.50 € - 808.02 €

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11380919
View Documents
Thermo Scientific Alfa Aesar
010681.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11390919
View Documents
Thermo Scientific Alfa Aesar
010681.09
10 g
196.98 €
10g
Please to purchase this item. Need a web account? Register with us today!
11370919
View Documents
Thermo Scientific Alfa Aesar
010681.18
50 g
808.02 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-75-5
78.08
WUKWITHWXAAZEY-UHFFFAOYSA-L
84512
[F-].[F-].[Ca++]
CaF2
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
CHEBI:35437

Specifications

∼2500°C
7789-75-5
Powder
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium difluoride
84512
78.08
Puratronic™
1.434
3.18 g/mL
Calcium fluoride
1403°C
White
CaF2
14,1667
Soluble in water (slightly).
[F-].[F-].[Ca++]
78.08
CHEBI:35437
99.985%
Odorless
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 232-188-7

missing translation for 'rtecsNumber' : EW1760000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only