Learn More
Calcium granules, redistilled, -16 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Appearance | Silvery-white granules |
Assay Percent Range | 99.5% (Metals basis) |
Quantity | 10 g |
Mesh Size | -16 Mesh |
Odor | Odorless |
Form | Granular |
Chemical Name or Material | Calcium, Redistilled |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z13K012 | Certificate of Analysis | 16/12/2024 | 042917.30,042917.18,042917.09 |
P25K003 | Certificate of Analysis | 05/04/2024 | 042917.30,042917.18,042917.09 |
Z15I015 | Certificate of Analysis | 25/01/2023 | 042917.30,042918.NB,042917.18,042917.09 |
M27I043 | Certificate of Analysis | 12/04/2022 | 042917.30,042918.NB,042917.18,042917.09 |
W17F067 | Certificate of Analysis | 12/04/2022 | 042917.30,042918.NB,042917.18,042917.09 |
Safety and Handling

- Calcium granules, redistilled
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- P223-Do not allow contact with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use carbon dioxide to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- MIXTURE LIST-Contains : Calcium
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.