missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hydride, 92% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 600°C (decomposition) |
Color | Gray |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | 92% min. |
Solubility Information | Decomposes in water,alcohol and acids to H2Soluble in water and alcohol. Insoluble in benzene. |
Formula Weight | 42.1 |
Sensitivity | Moisture sensitive |
Appearance | Light gray solid |
Density | 1.9 g/cm3 at 20°C |
Show More |
Safety and Handling
Product Identifier
- Calcium hydride
- Danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH014-Reacts violently with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Calcium hydride
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'