missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, Thermo Scientific Chemicals

96.58 € - 343.71 €

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10491431
SDS View Documents
Thermo Scientific Acros
199901000
100 g
96.58 €
100g
Please to purchase this item. Need a web account? Register with us today!
10542221
SDS View Documents
Thermo Scientific Acros
199905000
500 g
343.71 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-78-8
CaH2
42.09
MFCD00010897

Specifications

675.0°C
92% min. (Total Ca)
CaH2
01,105
Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
42.09
Extra Pure
1.9
Calcium hydride, extra pure, 0-2 mm grain size
7789-78-8
CaH2
MFCD00010897
15,1674
42.09
∼93%
Plastic bottle
1.9000g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.

GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF ON S

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-189-2

Product Identifier
  • Calcium hydride

Signal Word
  • Danger

Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.