missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 98%, extra pure

Product Code. 10637192 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10637192

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
SMILES [OH-].[OH-].[Ca++]

Specifications

Boiling Point 2850.0°C
Melting Point 580.0°C
Color White
Physical Form Amorphous Powder
Quantity 100 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Ca(OH)2
Merck Index 15, 1675
Solubility Information Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
IUPAC Name calcium;dihydroxide
Formula Weight 74.09
Percent Purity 98%
Grade Extra Pure
Identification Passes Tests
Packaging Plastic bottle
Density 2.24g/mL
Chemical Name or Material Calcium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.