missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, purum p.a., Honeywell™

Purum p.a.,≥96.0% (T)

80.86 € - 166.54 €

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15621940
SDS View Documents
Honeywell Research Chemicals
21181-1KG
1 kg
80.86 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15631940
SDS View Documents
Honeywell Research Chemicals
21181-5KG
5 kg
166.54 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

1305-62-0
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
6093208
calcium;dihydroxide
CaH2O2
MFCD00010901
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
CHEBI:31341
[OH-].[OH-].[Ca++]

Specifications

1305-62-0
Ca(OH)2
NONH for all modes of transport
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium;dihydroxide
6093208
74.09g/mol
purum p.a.
Calcium Hydroxide
CaH2O2
MFCD00010901
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
[OH-].[OH-].[Ca++]
74.09
CHEBI:31341
≥96% (T)
2.24g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

P261-P280-P305 + P351 + P338

H315-H318-H335

missing translation for 'einecsNumber' : 215-137-3

missing translation for 'rtecsNumber' : EW2800000

Product Identifier
  • Calcium hydroxide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Calcium dihydroxide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.