missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, Reag. Ph. Eur., puriss. p.a., Honeywell Fluka™
Puriss. p.a., Reag. Ph. Eur.,≥96%
39.95 € - 1989.82 €
Chemical Identifiers
CAS | 1305-62-0 |
---|---|
Molecular Formula | CaH2O2 |
Molecular Weight (g/mol) | 74.09 |
MDL Number | MFCD00010901 |
InChI Key | AXCZMVOFGPJBDE-UHFFFAOYSA-L |
Synonym | calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid |
PubChem CID | 6093208 |
ChEBI | CHEBI:31341 |
IUPAC Name | calcium;dihydroxide |
SMILES | [OH-].[OH-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15613300
|
Honeywell-Fluka
31219-100G |
100 g |
39.95 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643300
|
Honeywell-Fluka
31219-500G |
500 g |
63.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15623300
|
Honeywell-Fluka
31219-1KG |
1 kg |
95.30 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653300
|
Honeywell-Fluka
31219-6X1KG |
6 x 1 kg |
571.39 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15633300
|
Honeywell-Fluka
31219-25KG |
25 kg |
1989.82 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1305-62-0 | |
74.09 | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
6093208 | |
calcium;dihydroxide |
CaH2O2 | |
MFCD00010901 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
CHEBI:31341 | |
[OH-].[OH-].[Ca++] |
Specifications
1305-62-0 | |
Ca(OH)2 | |
NONH for all modes of transport | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
calcium;dihydroxide | |
6093208 | |
74.09g/mol | |
puriss. p.a. | |
Calcium Hydroxide, Reag. Ph. Eur. |
CaH2O2 | |
MFCD00010901 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
[OH-].[OH-].[Ca++] | |
74.09 | |
CHEBI:31341 | |
≥96% | |
2.24g/mL at 25°C |
Safety and Handling
P261-P280-P305 + P351 + P338
H315-H318-H335
missing translation for 'einecsNumber' : 215-137-3
missing translation for 'rtecsNumber' : EW2800000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium hydroxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Calcium dihydroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title