missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hypochlorite, Technical, Fisher Chemical™

Product Code. 10673933
Click to view available options
Quantity:
500 g
1 kg

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.98
MDL Number 10900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
IUPAC Name calcium dihypochlorite
SMILES [Ca++].[O-]Cl.[O-]Cl

Specifications

Melting Point 100°C
Color White
pH 11.4
Physical Form Solid
Quantity 1 kg
Formula Weight 142.98g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Calcium hypochlorite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H272-May intensify fire; oxidiser.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.