missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodate monohydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11339287
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium iodate monohydrate is used as oxidant added to lotions and ointments, acts as antiseptic and deodorant. It is used in the manufacture of disinfectants and iodine supplement in animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-32-0
Molecular Formula Ca(IO3)2·H2O
MDL Number MFCD00036265

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range >99%
Solubility Information Soluble in nitric acid and slightly soluble in water. Insoluble in ethanol.
Formula Weight 407.90 (389.88 Anhydrous)
Appearance Off-white powder
Chemical Name or Material Calcium iodate monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.