missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11348037
Click to view available options
Quantity:
25 g
100 g

Light Sensitive and Hygroscopic

Calcium iodide, anhydrous is used as a drug in upper respiratory combinations, expectorants. And it is also used as Laboratory chemicals, Manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]

Specifications

Boiling Point 1100°C
Melting Point 784°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (Metals basis)
Merck Index 14,1677
Solubility Information Freely soluble in water,methanol,ethanol,acetone; Insoluble in ether
Formula Weight 293.89
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 3.956 g/mL
Assay (metals basis)
Chemical Name or Material Calcium iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'