missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium iodide hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Calcium iodide hydrate used in photography. It is also used in cat food as a source of iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 42°C (decomposition) |
Color | Yellow |
Physical Form | Crystalline |
Quantity | 5 g |
Merck Index | 14,1677 |
Solubility Information | Freely soluble in water and alcohol |
Formula Weight | 293.89 (Anhydrous) |
Percent Purity | 99.9965% |
Grade | Puratronic™ |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'