missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium iodide, ultra dry, 99.99% (metals basis)
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
Calcium iodide is used in photography. It is also used in cat food as a source of iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Boiling Point | 1100°C |
| Melting Point | 784°C |
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.99% (Metal basis) |
| Solubility Information | Soluble in water,methanol,ethanol and acetone. Insoluble in ether. |
| Formula Weight | 293.89 |
| Sensitivity | Hygroscopic; Light sensitive |
| Appearance | Off-white to pale yellow powder or beads |
| Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu