missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.99% (metals basis)

Product Code. p-4407335
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11315907

Brand: Thermo Scientific Alfa Aesar 043674.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 10102-68-8 | CaI2 | 293.887 g/mol

Calcium iodide is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]

Specifications

Boiling Point 1100°C
Melting Point 784°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,methanol,ethanol and acetone. Insoluble in ether.
Formula Weight 293.89
Sensitivity Hygroscopic; Light sensitive
Appearance Off-white to pale yellow powder or beads
Density 3.95 g/cm3 at 20°C
Chemical Name or Material Calcium iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.