missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Lactate Pentahydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Specifications
Melting Point | 240°C |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 308.29g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Calcium lactate pentahydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction