missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11320929
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35054-52-5
Molecular Formula CaN2O6
Molecular Weight (g/mol) 164.09
MDL Number MFCD00149604
InChI Key ZCCIPPOKBCJFDN-UHFFFAOYSA-N
Synonym calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci
PubChem CID 16211464
SMILES [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1454
Merck Index 14,1682
Solubility Information Soluble in water.
IUPAC Name calcium;dinitrate;hydrate
Formula Weight 164.09 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic™
Sensitivity Hygroscopic
Density 1.82 g/mL
Assay (metals basis)
Chemical Name or Material Calcium nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'