Learn More
Calcium nitrate tetrahydrate, 99.98% (metals basis), Thermo Scientific Chemicals
Description
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 132°C (decomposition) |
Melting Point | 42°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99.98% |
Solubility Information | Soluble in water,alcohol and acetone. |
Formula Weight | 236.15 (164.09 Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling


- Calcium nitrate tetrahydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Calcium nitrate tetrahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.