Learn More
Calcium nitrate tetrahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
20.10 € - 151.00 €
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaN2O6·4H2O |
Molecular Weight (g/mol) | 236.15 |
MDL Number | MFCD00149604 |
InChI Key | ICSSIKVYVJQJND-UHFFFAOYSA-N |
Synonym | calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate |
PubChem CID | 16211656 |
ChEBI | CHEBI:86159 |
IUPAC Name | calcium;dinitrate;tetrahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10433171
|
Thermo Scientific Acros
423530250 |
25 g | Plastic bottle |
20.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731452
|
Thermo Scientific Acros
423535000 |
500 g | Plastic bottle |
46.60 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10494315
|
Thermo Scientific Acros
423530025 |
2.5 kg | Plastic bottle |
151.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13477-34-4 | |
236.15 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaN2O6·4H2O | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Specifications
132.0°C | |
13477-34-4 | |
5.0 to 7.0 (5% soln. at 25°C) | |
99.0 to 103% (Complexometry) | |
Ca(NO3)2·4H2O | |
15, 1684 | |
Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.15 | |
CHEBI:86159 | |
≥99% | |
1.86 | |
Calcium nitrate tetrahydrate |
45.0°C | |
White | |
Crystals | |
CaN2O6·4H2O | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 | |
ACS Reagent | |
1.8600g/mL |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : EW3000000
- Calcium nitrate tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only