missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, 99%, for analysis

Product Code. p-3734699 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 10733221

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132.0°C
Melting Point 45.0°C
Color White
pH 4.5 to 7.0 (5% soln. at 20°C)
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ca(NO3)2·4H2O
Merck Index 15, 1684
Solubility Information Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 236.15
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.86
Density 1.8600g/mL
Chemical Name or Material Calcium nitrate tetrahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium nitrate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.