Learn More
Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths), Thermo Scientific Chemicals
63.82 € - 215.07 €
Chemical Identifiers
CAS | 5794-28-5 |
---|---|
Molecular Formula | C2H2CaO5 |
Molecular Weight (g/mol) | 146.11 |
MDL Number | MFCD00150039 |
InChI Key | LQHWSGSWNOHVHO-UHFFFAOYSA-L |
Synonym | calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? |
PubChem CID | 165350 |
IUPAC Name | calcium;oxalate;hydrate |
SMILES | O.[Ca++].[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322108
|
Thermo Scientific Alfa Aesar
013007.06 |
5 g |
63.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332108
|
Thermo Scientific Alfa Aesar
013007.14 |
25 g |
215.07 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5794-28-5 | |
146.11 | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
165350 | |
O.[Ca++].[O-]C(=O)C([O-])=O |
C2H2CaO5 | |
MFCD00150039 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
calcium;oxalate;hydrate |
Specifications
200°C | |
White | |
99.9985% (Metal basis) | |
MFCD00150039 | |
Slightly soluble in water. | |
O.[Ca++].[O-]C(=O)C([O-])=O | |
146.11 | |
146.11 (128.10 Anhydrous) | |
Hygroscopic | |
2.12 g/cm3 |
5794-28-5 | |
Granules, Powder | |
C2H2CaO5 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
calcium;oxalate;hydrate | |
165350 | |
Odorless | |
White powder | |
Calcium oxalate monohydrate, H2O at 200° |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 209-260-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only