missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, 97+%, for analysis, powder, Thermo Scientific Chemicals

Product Code. 10381351
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium oxide, 97+%, CaO, CAS Number-1305-78-8, gebrannter kalk, calxyl, calcia, burnt lime, calcium oxide, airlock, calcium monoxide, calx, unslaked lime, 1kg, 2.0% max., 97% min. (CaO) on dry substance (Complexometry), 2850 deg.C, CHEBI:31344, White or Yellow, 215-138-9, 56.08, Analysis

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2570°C
Color White or Yellow
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97% min. (CaO) on dry substance (Complexometry)
Linear Formula CaO
Merck Index 15,1688
Solubility Information Solubility in water: reacts. Other solubilities: soluble in acids (reacts),soluble in glycerol,insoluble in alcohol
IUPAC Name calcium oxidandiide
Formula Weight 56.08
Percent Purity ≥97%
Grade Analytical
Loss on Ignition 5% max.
Packaging Plastic bottle
Chemical Name or Material Calcium oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium oxide, 97+%, for analysis, powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.