missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11391448
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in cement, glass, steel, paper, chemical and petroleum industries. It acts as a key ingredient in the process of cement making. It is widely used industrially as a dehydrating agent and an absorbent. Further it finds use as a water softener and as a pH regulator for wastewater. It is useful to detect traces of water in hydrocarbons, and in flue-gas desulfurization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2572°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.95% (Metal basis)
Solubility Information Slightly soluble in water. Insoluble in methanol
IUPAC Name oxocalcium
Formula Weight 56.08
Odor Odorless
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Appearance White powder
Density 3.37
Chemical Name or Material Calcium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.