missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Oxide, Honeywell Fluka™
Puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)
38.88 € - 224.35 €
Chemical Identifiers
CAS | 1305-78-8 |
---|---|
Molecular Formula | CaO |
Molecular Weight (g/mol) | 56.08 |
MDL Number | MFCD00010911 |
InChI Key | BRPQOXSCLDDYGP-UHFFFAOYSA-N |
Synonym | calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx |
PubChem CID | 14778 |
ChEBI | CHEBI:31344 |
SMILES | [O--].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15681090
|
Honeywell-Fluka
12047-1KG |
1 kg |
38.88 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691090
|
Honeywell-Fluka
12047-2.5KG |
2.5 kg |
66.18 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15611100
|
Honeywell-Fluka
12047-5KG |
5 kg | N/A | ||||||
15621100
|
Honeywell-Fluka
12047-6X1KG |
6 x 1 kg |
224.35 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601100
|
Honeywell Fluka
12047-20KG |
20 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1305-78-8 | |
56.08 | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
14778 | |
[O--].[Ca++] |
CaO | |
MFCD00010911 | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
CHEBI:31344 |
Specifications
1305-78-8 | |
CaO | |
MFCD00010911 | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
[O--].[Ca++] | |
56.08 | |
CHEBI:31344 | |
96 to 100.5% (ex ignited substance) | |
3.3g/mL at 25°C |
Fine Powder | |
CaO | |
1910 | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
calcium oxidandiide | |
14778 | |
56.08g/mol | |
puriss. | |
Calcium Oxide, Meets analytical specification of FCC |
Safety and Handling
P261-P280-P305 + P351 + P338
H315-H318-H335
missing translation for 'einecsNumber' : 215-138-9
missing translation for 'rtecsNumber' : EW3100000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium oxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Calcium oxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title