missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Pantothenate, U.S.P., J.T. Baker™
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Quantity | 25 g |
Formula Weight | 476.54 |
Grade | USP |
Packaging | Wide Mouth Amber Glass Bottle |
Chemical Name or Material | Calcium Pantothenate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction