missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, 96%, pure, mixture of Calcium phosphates, Thermo Scientific Chemicals

Product Code. 12075390
Click to view available options
Quantity:
1 kg
5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.18
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670.0°C
Quantity 5 kg
Assay Percent Range 95% min. (Complexometry)
Linear Formula Ca3(PO4)2
Merck Index 15,1696
Solubility Information Solubility in water: 0.1g/L (25°C)
Formula Weight 310.18
Percent Purity 96%
Grade Pure
Packaging Plastic drum
Chemical Name or Material Calcium phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'