Learn More
Calcium phosphate, dibasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
16.98 € - 348.74 €
Chemical Identifiers
CAS | 7757-93-9 |
---|---|
Molecular Formula | CaHO4P |
Molecular Weight (g/mol) | 136.06 |
MDL Number | MFCD00010909 |
InChI Key | FUFJGUQYACFECW-UHFFFAOYSA-L |
Synonym | calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1 |
PubChem CID | 24441 |
ChEBI | CHEBI:32596 |
IUPAC Name | calcium;hydrogen phosphate |
SMILES | [Ca++].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10318400
|
Thermo Scientific Acros
215710250 |
25 g | Plastic bottle |
16.98 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10770911
|
Thermo Scientific Acros
215710010 |
1 kg | Plastic bottle |
109.55 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11375922
|
Thermo Scientific Acros
215710050 |
5 kg | Plastic Drum |
348.74 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7757-93-9 | |
136.06 | |
FUFJGUQYACFECW-UHFFFAOYSA-L | |
24441 | |
calcium;hydrogen phosphate |
CaHO4P | |
MFCD00010909 | |
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1 | |
CHEBI:32596 | |
[Ca++].OP([O-])([O-])=O |
Specifications
7757-93-9 | |
Powder | |
CaHO4P | |
MFCD00010909 | |
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1 | |
FUFJGUQYACFECW-UHFFFAOYSA-L | |
calcium;hydrogen phosphate | |
24441 | |
136.06 | |
Pure |
White | |
96% min. (Complexometry) | |
CaHPO4 | |
15,1694 | |
Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol | |
[Ca++].OP([O-])([O-])=O | |
136.06 | |
CHEBI:32596 | |
97% | |
Calcium phosphate, dibasic |
Safety and Handling
missing translation for 'einecsNumber' : 231-826-1
- Calcium phosphate, dibasic
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.