missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals

26.93 € - 85.02 €

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12675027
View Documents Promotion Details
Thermo Scientific Acros
446430500
50 g Plastic bottle
26.93 €
50g
Please to purchase this item. Need a web account? Register with us today!
12665027
View Documents Promotion Details
Thermo Scientific Acros
446432500
250 g Plastic bottle
85.02 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

7757-93-9
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
24441
calcium;hydrogen phosphate
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
CHEBI:32596
[Ca++].OP([O-])([O-])=O

Specifications

7757-93-9
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium;hydrogen phosphate
24441
136.06
Analytical
98.0 to 103% (Complexometry)
CaHPO4
15,1694
Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
[Ca++].OP([O-])([O-])=O
136.06
CHEBI:32596
≥98%
Calcium phosphate, dibasic
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-826-1

Product Identifier
  • Calcium phosphate, dibasic

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only